missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127 / IL-7Ra Rat, V450, Clone: A7R34, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB55-V450-25ug
This item is not returnable.
View return policy
Specifications
CD127 / IL-7Ra | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Il17ra | |
Protein A/G Purified | |
RUO | |
16172 | |
IgG2a κ |
Flow Cytometry | |
A7R34 | |
Violet Fluor 450 (V450) | |
NP_032385 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction