Learn More
CD178 / Fas ligand Mouse, Biotin, Clone: NOK-1, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio QAB85-B-100ug
 View more versions of this product
Product Code. 15942568
  • 312.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
CD178 / Fas ligand
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
FASLG
Protein A/G Purified
RUO
356
IgG1 κ
Flow Cytometry
NOK-1
Biotin
NP_001289675
Mouse
100 μg
Primary
Store at 2-4°C.