missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD234 (DARC) Mouse anti-Human, PE, Clone: NaM185-2C3 (also known as 2C3), BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566424
This item is not returnable.
View return policy
Description
CD234 (DARC) Monoclonal antibody specifically detects CD234 (DARC) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD234 (DARC) | |
| Monoclonal | |
| PE | |
| 100 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
| Flow Cytometry | |
| NaM185-2C3 (also known as 2C3) | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction