missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat, APC, Clone: RM4-5, enQuireBio™
Rat Monoclonal Antibody
98.45 € - 225.00 €
Specifications
Antigen | CD4 |
---|---|
Clone | RM4-5 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952608
|
EnquireBio
QAB9-APC-25ug |
25 μg |
98.45 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942608
|
EnquireBio
QAB9-APC-100ug |
100 μg |
225.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD4 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
12504 | |
IgG2a κ | |
Protein A/G Purified |
RM4-5 | |
Flow Cytometry | |
APC | |
RUO | |
NP_038516 | |
Cd4 | |
Primary | |
Store at 2-4°C. |