missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat, V450, Clone: GK1.5, enQuireBio™
Rat Monoclonal Antibody
137.00 € - 301.00 €
Specifications
Antigen | CD4 |
---|---|
Clone | GK1.5 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962528
|
EnquireBio
QAB8-V450-25ug |
25 μg |
137.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15952528
|
EnquireBio
QAB8-V450-100ug |
100 μg |
301.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD4 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
12504 | |
IgG2b κ | |
Protein A/G Purified |
GK1.5 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
NP_038516 | |
Cd4 | |
Primary | |
Store at 2-4°C. |