missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD42b/GPIb alpha Antibody (486805), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4067W
This item is not returnable.
View return policy
Description
CD42b/GPIb alpha Monoclonal antibody specifically detects CD42b/GPIb alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CD42b/GPIb alpha | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 486805 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
| Mouse myeloma cell line NS0-derived recombinant human CD42b/GPIb alpha , His17-Leu505, Accession # P07359 | |
| 0.1 mL | |
| Immunology | |
| 2811 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction