Learn More
CD79a Mouse anti-Human, PE, Clone: HM47, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555935
239.68 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The HM47 monoclonal antibody specifically binds to the cytoplasmic domain of CD79a. CD79a is a 47 kDa type 1 transmembrane glycoprotein present on B lymphocytes. CD79a is also referred to as mb-1, IGA and Ig-alpha (Ig α ). It is expressed on B cells at various stages of differentiation, from pre-B cell stage, probably before expression of cytoplasmic µ chain, to plasma cell stage, in which it is detected only in the cytoplasm. CD79a associates with CD79b to form part of the B-cell receptor complex. It has been suggested that CD79a may play a role in mediating the transport of IgM to the cell surface. This antibody has been found to crossreact with permeabilized A20 cells (mouse B cell line).

Specifications
CD79a | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD79a Peptide | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
HM47 | |
CD79a, immunoglobulin-associated alpha ; CD79A; Ig-alpha; IGA; Igα; MB-1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.