Learn More
CD8 Mouse anti-Human, Unconjugated, Clone: 1G2B10, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | CD8 |
|---|---|
| Clone | 1G2B10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16829135
|
Proteintech
66868-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16819135
|
Proteintech
66868-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD8. This antibody recognizes human antigen. The CD8 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| CD8 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 925 | |
| Primary |
| 1G2B10 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.