Learn More
STEMCELL Technologies CellAdhere™ Dilution Buffer
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- Vitronectin XF™, developed and manufactured by Nucleus Biologics, is an effective alternative to Corning™ Matrigel™
- Vitronectin XF™ is a defined, xeno-free cell culture matrix that supports the growth and differentiation of human pluripotent stem cells
- When used with mTeSR™1, mTeSR™ Plus, TeSR™-E8™, or TeSR™2, Vitronectin XF™ provides a defined culture system for the maintenance of human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells under feeder-free conditions
- This system allows complete control over the culture environment, resulting in more consistent cell populations and reproducible results in downstream applications
- Note: Catalog #07180 is the matrix only and requires additional reagents (Catalog #07183, 07174 or 05872, and 27147) available for purchase separately or in a kit with Vitronectin XF™ (Catalog #07190 or 07191)
Specifications
Specifications
| Description | Endoderm, PSC-Derived; Pluripotent Stem Cells |
| Quantity | 100 mL |