missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Poly-D-lysine Coated Cell Culture Dish
For cultivation in serum-free and serum reduced medium
130.00 € - 804.00 €
Specifications
Closure Color | Clear |
---|---|
Closure Included | Closure Included |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Color | Clear |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
11729442
|
Greiner Bio-One
664940 |
100 mm |
490.00 €
Pack of 40 |
Estimated Shipment: 17-03-2025
Log in to see stock. |
||||||
10756542
|
Greiner Bio-One
664940 |
100 mm | N/A | |||||||
10480332
|
Greiner Bio-One
628940 |
60 mm | N/A | |||||||
11749442
|
Greiner Bio-One
628940 |
60 mm |
804.00 €
Pack of 100 |
Estimated Shipment: 17-03-2025
Log in to see stock. |
||||||
Description
Greiner Bio-One CELLCOAT Poly-D-lysine Coated Cell Culture Vessel is used for cultivation of transfected cells and fastidious cell lines, as well as serum-free or -reduced cell culture.
- Source: Synthetic
- Cultivation of neuronal cells, transfected cells and other fastidious cell lines
- Cultivation in serum-free and serumreduced medium
- Improvement of cell attachment
Specifications
Clear | |
Polystyrene | |
Clear | |
Round | |
Poly-D-Lysine | |
Lid |
Closure Included | |
Flat | |
Polystyrene | |
Non-sterile | |
Cell Culture Dish |
For research use only. Not for use in diagnostic procedures, unless otherwise indicated
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One CELLCOAT™ Poly-D-lysine Coated Cell Culture Dish