Learn More
Corning™ CellCube™ Modules
BIOPHARMAGREENER_CHOICE
Product Code. p-7221811 Shop All Corning Products
Click to view available options
Capacity (Metric):
600 mL
1.5 L
Tray Layers:
10
25
Surface Treatment:
CellBIND
Tissue Culture-treated
missing translation for 'unitSize'
Each
Pack of 2
Product Code. 17279099
missing translation for 'mfr': Corning™ 3200
Only null left
Add to basket
Add to basket
The CellCube™ module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth.
CellCube Modules provide:
  • a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
  • polystyrene growth surface optimally treated for the growth of attachment dependent cells.
  • a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen.
  • an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
TRUSTED_SUSTAINABILITY
Specifications
Description Individually bagged
Capacity (Metric) 600 mL
Graduations No
No. per Case 2
No. per Pack 1
Product Type CellCube Module
Shape Square
Sterility Sterile
Culture Area 8500 cm2
Surface Treatment Tissue Culture-treated
Tray Layers 10
Sterilization Method Yes