missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellMask™ Orange Actin Tracking Stain

Stain and track intracellular F-actin with CellMask Orange Actin Tracking Stain in live or fixed cells in ICC, IHC, and 3D cell culture. The stain easily permeates live-cell membranes, is retained within the cells after loading, and shows no detectable effects on viability after 24 hours of incubation.

Brand:  Invitrogen™ A57247

 View more versions of this product

Product Code. 17148568

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Stain and track intracellular F-actin with CellMask Orange Actin Tracking Stain in live or fixed cells in immunofluorescence (IF), immunocytochemistry (ICC), and immunohistochemistry (IHC) protocols, and in 3D cell culture. The CellMask Orange Actin Tracking Stain fluorescently labels polymerized/filamentous actin (F-actin) in live or fixed cells because it is designed to readily permeate live cells, resulting in bright and uniform labeling. CellMask Orange Actin Tracking Stain can be used to track F-actin in live cells for 24 hours or more. Live cells stained with CellMask Orange Actin Tracking Stain can be fixed; the stain can also be used on already fixed cells. Cell Mask Orange Actin Tracking Stain can be multiplexed with antibodies in IF, ICC, and IHC protocols, and can also be used in 3D cell culture for 3D imaging.

Features of Cell Mask Orange Actin Tracking Stain include the following:
• Stains F-actin in live or fixed cells for fluorescence 2D monolayer or 3D imaging experiments
• Can be multiplexed with antibodies in IF/ICC/IHC experiments
• Fluorescence maxima (Excitation/Emission): 545/570 nm; can be detected with TRITC/RFP filter

CellMask actin tracking stains are available in three colors: green (Cat. No. A57243, A57246), detected with a FITC/GFP filter (Ex 503 nm/Em 512 nm), orange (Cat. No. A57244, A57247) detected with a TRITC/RFP filter (Ex 545 nm/Em 570 nm), and deep red (Cat. No. A57245, A57248), detected with a CY5/Deep Red filter (Ex 652 nm/Em 669 nm).

CellMask actin tracking stains detect F-actin in live or fixed cells or tissue with a targeting structure that closely resembles jasplakinolide. They do not label globular actin (G-actin) or monomer actin (M-actin). CellMask actin tracking stains easily permeate live cell membranes and are retained within live cells after loading. As demonstrated in the figures below, incubation with CellMask actin tracking stains has no detectable effect on viability of various cell lines after 24 hours of incubation. In addition, CellMask actin tracking stains can be used to detect F-Actin in paraformaldehyde fixed cells or tissue (freshly fixed or cryo frozen only) and can be multiplexed with antibody detection.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orange
570 nm
Fluorescence Microscope, High Content Analyzer
Fluorescence Microscopy
545 nm
Room Temperature
Fluorescent Dye
Actin, Cytoskeleton
Contains 5 vials with enough reagent to stain 600 slides, when 500 μL of 1X staining solution is used per slide.

• Add 60 μL/vial of anhydrous DMSO to make a 1000X (1 mM) stock solution
• Dissolve stock solution 1:1000 in cell imaging solution to make 1X (1 μM) staining solution

Store at room temperature. Can be stored to –20°C with no freeze/thaw cycles. When stored as instructed, these reagents are effective for a minimum of 6 months after receiving.

TRITC/RFP
5 x 60 Assays
CellMask™ Orange Actin Tracking Stain
CellMask™
F-Actin
Actin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.