missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow are recommended for detection of yeasts and molds, particle count, and microscopy.
Brand: Sartorius Stedim Biotech™ 130H6--50----ACN
This item is not returnable.
View return policy
Description
- Sterile, individually packaged filters
- Save preparatory time
- Perfect grid printing
- Filter colors allow the best contrast to the colonies or particles
- Outstanding microorganism recovery rate
- High flow membranes
- Sterilized by gamma irradiation
- Compatible with aqueous solutions (pH 4-8), hydrocarbons and several other organic solvents
Specifications
Gray/White Grid | |
0.45 μm | |
130 | |
115 to 145 μm |
Cellulose Nitrate (Cellulose Mixed Ester) | |
Sterile | |
100/pk. | |
50 mm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- X100 Cellulose Nitrate Grey - White_ Sterile_ 0,45
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose