Learn More
B Braun™ Cellulose Absorbent Pads
Cellulose swabs off the roll, made of highly bleached cellulose dressing according to DAB, DIN 19310. For skin cleansing before injections, as an absorbent pad. Can be torn off easily and cleanly. Swab size: 4 x 5 cm, punched into ready-to-use shape.
Brand: B Braun™ 9051015
Description
- Non-sterile
- Perforated design
Your input is important to us. Please complete this form to provide feedback related to the content on this product.