Learn More
Abnova™ CEN9 (Orange) FISH Probe
Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.
Brand: Abnova™ FC0192.200uL
This item is not returnable.
View return policy
Specifications
Store at -20°C in the dark. | |
FISH | |
200 μL |