missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103489
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CENPC1.
Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.
Specifications
| CENPC | |
| Polyclonal | |
| Unconjugated | |
| CENPC | |
| CENPC; CENP-C; CENP-C 1; Cenpc1; Centromere autoantigen C; centromere autoantigen C1; centromere protein C; CENTROMERE PROTEIN C (CENP-C) (CENTROMERE AUTOANTIGEN C); centromere protein C 1; centromere protein C1; hcp-4; ICEN7; Interphase centromere complex protein 7; MIF2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1060 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q03188 | |
| CENPC | |
| A synthesized peptide derived from human CENPC(Accession Q03188), corresponding to amino acid residues K797-Y847. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction