Learn More
BINDER™ Temperature Calibration Certificate with 1 Measurement Point
For Use With: BINDER units B 23
Brand:  BINDER™ 8012-0030
Product Code. 15691646
  • 472.00 € / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
Specifications
Calibration certificate, temperature - for temperature, measurement in center of chamber at specified temperature
BINDER Chambers B 23
Temperature Calibration Certificate
Temperature Measurement
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Temperature Calibration Certificate with 1 Measurement Point > Calibration Points: 1 Measurement Point, For Use With: All ED, FD, FED, FP, M, MK, BD, BF, KB, KBW models, and FDL 115, MDL 115