Learn More
OHAUS™ Centrifuge Adapter
Shop All Ohaus Products
Click to view available options
Capacity:
15 x 1.5/2.0 mL
175/225 mL
250 mL
3 x 50 mL
4 x 30 mL
50 mL
6 x 15 mL
Description:
FA x2
FA x3
FB x2
MTB x2
RB x2
Bore Size (Metric):
Depth 11 mm
Depth 17 mm
Depth 25.5 mm
Depth 29 mm
Depth 29.5 mm
Depth 60 mm
Depth 60.5 mm
Depth 61 mm
Depth 61.5 mm
missing translation for 'unitSize'
Pack of 2
Pack of 3
Description
- Adapter for various Centrifuge Multi models
- Various capacities available
Specifications
Specifications
| Type | Adapter |
| Capacity | 175/225 mL |
| For Use With (Equipment) | Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V |
| Bore Size (Metric) | Depth 61.5 mm |
| Description | FA x2 |