Learn More
Karl Hecht™ CERAN™ Laboratory Protective Plate
Product Code. 10680772
Click to view available options
Dimensions (L x W):
155 x 155 mm
Unit Size:
Each
Product Code. 10680772
Brand: Karl Hecht™ 41066002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from CERAN™ glass ceramic.
  • 155 x 155mm
  • Each in a cardboard bag
  • For general laboratory usage
Specifications
Product Type Protection Plate
Length (Metric) 155 mm
Material Glass Ceramic
Dimensions (L x W) 155 x 155 mm
For Use With (Application) General Laboratory Usage
Width (Metric) 155 mm