Learn More
CHAC1 Monoclonal Antibody (OTI4B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507048S
Description
CHAC1 Monoclonal antibody specifically detects CHAC1 in Human samples. It is validated for Western BlotSpecifications
| CHAC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q76N32 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23177 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4B3 | |
| Unconjugated | |
| CEP68 | |
| 1810008K03Rik; Blocks Notch protein; botch; cation transport regulator homolog 1; cation transport regulator-like protein 1; ChaC glutathione specific gamma-glutamylcyclotransferase 1; ChaC glutathione-specific gamma-glutamylcyclotransferase 1; ChaC, cation transport regulator 1; ChaC, cation transport regulator homolog 1; ChaC, cation transport regulator-like 1; Chac1; Gamma-GCG 1; gamma-GCT acting on glutathione homolog 1; glutathione-specific gamma-glutamylcyclotransferase 1; Neuroprotective protein 7; Npg7; RGD1307153 | |
| Full length human recombinant protein of human CEP68 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.