Learn More
Thermo Scientific™ Chemical Kit for TruScan G3/RM/GP
Product Code. 18021081 Shop All Thermo Scientific Products
Click to view available options
Type:
Certification Kit
missing translation for 'unitSize'
Each
Product Code. 18021081
missing translation for 'mfr': Thermo Scientific™ 5800150101
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Raman Spectrometer Certification Kit for the Analytical Instrument Qualification of TruScan, TruScan RM or TruScan GP. Kit to be used in conjunction with IQ/OQ/PQ document.
  • Certification kit validity is good for one year from date of manufacture
  • Kit consists of qualified sample lots of Polystyrene, Cyclohexane, and Acetaminophen
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ramina Process Analyzer
Type Certification Kit