missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Chemical Oxygen Demand (COD) Test Kits
Click to view available options
Range:
13 at 20°C
20 to 1500 mg/L
200 to 15,000 mg/L
25 to 1500 mg/L
300 to 3500 mg/L
4 to 40 mg/L
4 to 5 at 25°C
6 at 20°C
9 at 20°C
Unit Size:
0.75kg
136g
400mL
Pack of 25
Description
- Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
- Safe determinations of COD in water
- Chlorine dioxide (ClO<sub>2</sub>) reagent test kit
- For waste water, production control, ground water, surface water
- For PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, spectroFlex photometers
Specifications
Specifications
| Certifications/Compliance | EN ISO 15705 |
| For Use With (Application) | Wastewater, production control, ground water, surface water |
| For Use With (Equipment) | PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Ph |
| No. of Tests | 50 |
| Range | 13 at 20°C |
| Type | CombiCheck Lead (Pb) reagent test kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction