missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Chemical Oxygen Demand (COD) Test Kits

Get accurate Chemical Oxygen Demand (COD) test results with the Chlorine dioxide reagent test kit.

Brand:  WTW™ 252071


 View more versions of this product

Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10396363

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
  • Safe determinations of COD in water
  • Chlorine dioxide (ClO<sub>2</sub>) reagent test kit
  • For waste water, production control, ground water, surface water
  • For PhotoLab&reg; S6, PhotoLab&reg; S12, PhotoLab&reg; Spectral, spectroFlex photometers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN ISO 15705
Wastewater, production control, ground water, surface water
200
25 to 1500 mg/L
10 mL
PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Ph
1 Pack
Chlorine test kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • COD Cell Test C4/25 Method: photometric 25 - 1500 mg/l

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Carcinogenicity Category 1B

Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H290-May be corrosive to metals.
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, mercury(II) sulphate, potassium dichromate

SDS
Documents

Documents

Certificates
Promotions

Promotions