Learn More
Chlorite (ClO2-) Standard for Ion Chromatography, 1000 mg/L in NaOH 0.1%, Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Specifications
| Chemical Name or Material | Chlorite Standard for Ion Chromatography |
| Density | 1.01 g/cm3 |
| Quantity | 100 mL |
| Grade | Standards for Ion Chromatography |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.