missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, Extra Pure, SLR, Fisher Chemical™

Product Code. 10243660 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
This item is not returnable. View return policy

Product Code. 10243660

Brand: Fisher Chemical C/4680/PB17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.56
MDL Number 530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
IUPAC Name chlorobenzene
SMILES ClC1=CC=CC=C1

Specifications

Melting Point -45°C
Boiling Point 131°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 12mbar at 20°C
Viscosity 0.8 mPaS at 20°C
Formula Weight 112.56g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.