missing translation for 'onlineSavingsMsg'
Learn More

Chlorodifluoroacetic acid, 98+%, Thermo Scientific™

Product Code. 11406957
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a starting material for generating difluorocarbene. It is also used as a pharmaceutical and agrochemicals intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, bases, active metals and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-04-0
Molecular Formula C2HClF2O2
Molecular Weight (g/mol) 130.475
MDL Number MFCD00004176
InChI Key OAWAZQITIZDJRB-UHFFFAOYSA-N
Synonym chlorodifluoroacetic acid, acetic acid, chlorodifluoro, difluorochloroacetic acid, chloro difluoro acetic acid, 2-chloro-2,2-difluoro-acetic acid, acetic acid, 2-chloro-2,2-difluoro, snpljqdjptadtaetp@, chlorodifluoroaceticacid, chlorodifluoro acetic acid, acmc-209p1k
PubChem CID 66154
IUPAC Name 2-chloro-2,2-difluoroacetic acid
SMILES C(=O)(C(F)(F)Cl)O

Specifications

Melting Point 20°C to 24°C
Boiling Point 121°C to 122°C
Odor Acrid
Quantity 25 g
UN Number UN3265
Beilstein 956625
Formula Weight 130.48
Percent Purity ≥98%
Chemical Name or Material Chlorodifluoroacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'