Learn More
Azpack™ Matte Vellum Paper Labels
Adhere to a variety of surfaces. Azpack™ Matte Vellum Paper Labels can be used with a thermal transfer printer to create labels.
Brand: Azpack™ LAB-200-071P
This item is not returnable.
View return policy
Description
- Matte white vellum paper
- Thermal transfer
- Glassine finish
- Permanent adhesive backing
Specifications
Butt cut short edge | |
Vellum | |
64 x 25 mm |
White | |
500/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Azpack™ Matte Vellum Paper Labels > Dimensions: 64L x 25mmW
Spot an opportunity for improvement?Share a Content Correction