Learn More
The Central Surgical Company™ Chroma Gesellschaft™ Stainless Steel Hypodermic Needle
Click to view available options
Needle Gauge:
14 G
17 G
21 G
23 G
24 G
Length (Metric) Needle:
25 mm
38 mm
101 mm
152 mm
Unit Size:
Pack of 10
Specifications
Specifications
| Material | Stainless Steel |
| Needle Gauge | 17 G |
| Product Type | Hypodermic Needle |
| Length (Metric) Needle | 101 mm |