Learn More
Macherey-Nagel™ Chromabond™ BT 25 SiOH Column
Brand: Macherey-Nagel™ 732962
This item is not returnable.
View return policy
Specifications
165 mm | |
Flash Column | |
Biotage Isolera Systems |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Chromabond™ BT 25 SiOH Column > I.D.: 21.2mm
Spot an opportunity for improvement?Share a Content Correction