Learn More
Macherey-Nagel™ Chromabond™ Drug II Polypropylene SPE Column
Product Code. 12877891 Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
100 mg
200 mg
500 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
Unit Size:
Pack of 100
Pack of 50
Product Code. 12877891
Brand: Macherey-Nagel™ 730686
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Base Material Silica
Column Capacity (Metric) 3 mL
Column Type SPE
Bed Weight 500 mg
For Use With (Application) Solid Phase Extraction Analysis of THC and Derivatives Acidic Analytes From Biological Fluids
Particle Size 45 μm
Sorbent Chromabond Drug II
Type Solid Phase Extraction Column
Pore Size 60 Å
Surface Area (Metric) 500 m2/g