Learn More
Macherey-Nagel™ Chromabond™ NAN Polypropylene SPE Column
Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
400/1400/400 mg
700/2000/700 mg
Column Capacity (Metric):
3 mL
6 mL
Unit Size:
Pack of 30
Pack of 50
Specifications
Specifications
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| Bed Weight | 700/2000/700 mg |
| For Use With (Application) | Solid Phase Extraction Analysis of PCBs From Sludge |
| Sorbent | Chromabond NAN |
| Type | Solid Phase Extraction Column |