Learn More
Sartorius Stedim Biotech™ Chromatography Paper, Roll, Grade FN 100
Product Code. 15932720 Shop All Sartorius Products
Click to view available options
Dimensions (L x W):
270 mm x 100 m
Unit Size:
Each
Product Code. 15932720
Brand: Sartorius Stedim Biotech™ FT15270100100
Please to purchase this item. Need a web account? Register with us today!
Sartorius Stedim Biotech™ Chromatography Paper, Roll, Grade FN 100 is ideal for blotting, chromatography, and other absorption applications.
  • Highly pure paper grade.
  • Ideal for blotting and chromatography.
  • Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
TRUSTED_SUSTAINABILITY
Specifications
Description Non-sterile, bulk packed reel, white
Dimensions (L x W) 270 mm x 100 m
Film Thickness 0.35 mm
For Use With (Application) Chromatography, blotting
Grade FN 100
Material Cotton Linters