Learn More
Cilomilast, MedChemExpress
MedChemExpress Cilomilast (SB-207499) is a potent, selective and orally active inhibitor of Phosphodiesterase 4 (PDE4), with IC50s of ∼100 and 120 nM for LPDE4 and HPDE4, respectively.
Brand:  Medchem Express HY-10790
Encompass_Preferred
 View more versions of this product
Product Code. 16464158
  • 167.40 € / 10mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Cilomilast shows selectivity for PDE4 over PDE1, PDE2, PDE3 and PDE5 (IC50=74, 65, >100, and 83 μM, respectively). Cilomilast has anti-inflammatory and immunomodulatory effects and can be used for thr research of asthma and chronic obstructive pulmonary disease (COPD).
Specifications
Specifications
Cilomilast
White
C20H25NO4
SB-207499
O=C([C@H]1CC[C@@](C#N)(CC1)C2=CC=C(OC)C(OC3CCCC3)=C2)O
343.42
Research
COVID-19-immunoregulation
153259-65-5
Research
10 mg
DMSO : ≥ 100 mg/mL (291.19 mM)
343.42
99.89%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cilomilast, MedChemExpress