Learn More
Hirschmann™ Class A Amber Glass White Graduated Measuring Flasks
Provided in amber glass for the protection of light-sensitive contents.
Brand:  Hirschmann™ 264018527
 View more versions of this product
Product Code. 10652364
  • 102.00 € / Pack of 2
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • White graduation
  • Class A
  • With dated batch identification
  • Standard ground and PE stopper
  • Manufactured from DURAN™ borosilicate glass for high chemical and thermal resistance
  • USP certified
Specifications
Specifications
Polyethylene
200 mL
Liquid handling of light-sensitive products
Ground 14/23
A
Stopper
Amber
Borosilicate Glass
210 ± 5 mm
Yes
Flat