Learn More
Hirschmann™ Class B Graduated Cylinders with Blue Graduations
Product Code. 11671409
Click to view available options
Unit Size:
Pack of 12
Pack of 3
Pack of 6
Capacity (Metric):
500 mL
1000 mL
Product Code. 11671409
Brand: Hirschmann™ 7501194
Please to purchase this item. Need a web account? Register with us today!
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann™ Class B Measuring Cylinders, raised scale are manufactured from PP.
  • Acc. to DIN 12681/ISO 6706
  • PP
  • Raised scale
  • Blue graduated
  • Main point ring graduation
  • Hexagonal base
Specifications
Class B
Disposable Reusable
Shape Round
For Use With (Application) Precise Liquid Measurement and Fluid Transfer in the Laboratory
Scale Color Blue
Base Type Hexagonal
Height (Metric) 440 mm
Type Graduated Measuring Cylinder