Learn More
Hirschmann™ Class B Graduated Cylinders with Blue Graduations
Click to view available options
Unit Size:
Pack of 12
Pack of 3
Pack of 6
Capacity (Metric):
500 mL
1000 mL
Description
- Acc. to DIN 12681/ISO 6706
- PP
- Raised scale
- Blue graduated
- Main point ring graduation
- Hexagonal base
Specifications
Specifications
| Class | B |
| Disposable | Reusable |
| Shape | Round |
| For Use With (Application) | Precise Liquid Measurement and Fluid Transfer in the Laboratory |
| Scale Color | Blue |
| Base Type | Hexagonal |
| Height (Metric) | 200 mm |
| Type | Graduated Measuring Cylinder |