Learn More
Zwiebel™ Class M1 Weight with Certificate
Zwiebel™ M1 Weight with Certificate is intended for use in the verification or calibration of class M2 weights.
Brand: Zwiebel™ PBI2300Y
This item is not returnable.
View return policy
Description
- Permanent magnetization: < 250 μT
- 0.6 mm thickness
- One bloc material without adjustment cavity
- Weights intended for use in the verification or calibration of class M weights, and for use with weighing 2 instruments of medium accuracy class III
Specifications
Aluminum Sheet | |
500 mg | |
Pentagonal | |
13.5 mm |
Recommendation OIML R111 | |
2700 ±100 kg/m3 | |
20.7 mm | |
Certificate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zwiebel™ Class M1 Weight with Certificate > Weight: 500 mg
Spot an opportunity for improvement?Share a Content Correction