missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M1 Weight Set with Certificate

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.

137.00 € - 145.00 €

Specifications

For Use With (Application) For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
View More Specs

Products 2
Product Code Brand Mass Price Quantity & Availability  
Product Code Brand Mass Price Quantity & Availability  
11890922
View Documents
Zwiebel™
PBI5682X
1 to 500 mg
137.00 €
Each
Estimated Shipment: 18-12-2024
to see stock.
Add to basket
 
11870932
View Documents
Zwiebel™
PBI56836
1 to 50 g
145.00 €
Each
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
 
Description

Description

  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications

    Specifications

    For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
    Videos
    SDS
    Documents

    Documents

    Product Certifications