missing translation for 'onlineSavingsMsg'
Learn More

Clathrin, Light Chain Mouse, Clone: CLC/1421, enQuireBio™

Mouse Monoclonal Antibody

199.00 € - 528.00 €

Specifications

Antigen Clathrin, Light Chain
Clone CLC/1421
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15939817
View Documents
EnquireBio
1211-MSM1-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15949817
View Documents
EnquireBio
1211-MSM1-P1
100 μg
528.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Clathrin, Light Chain
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
1212
Purified recombinant N-terminal fragment of human Clathrin Light Chain
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
31-44kDa
CLC/1421
Immunohistochemistry
Unconjugated
RUO
XP_016864515
CLTA and CLTB
IgG2b κ
Protein A/G Purified
Cytoplasmic
HeLa cells. Placenta or Prostate Carcinoma.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.