Learn More
Scotch-Brite™ Clean & Shine Floor Pad
This 2 in 1 daily cleaning pad for low speed scrubbers gradually increases shine with repeated use
Brand: Scotch-Brite™ 7100148105
This item is not returnable.
View return policy
Description
- Cleans and shines in the same step, using only a low-speed scrubber
- Save costs and labor by reducing or eliminating the need to burnish
- Removes black marks 3 times faster than traditional cleaning pads
- Two-sided pad lasts longer than traditional cleaning pads
- Effective on most coated and uncoated hard floors including VCT, vinyl, rubber, marble, stone, terrazzo, granite, and concrete with only water or neutral cleaner
Specifications
Cleaning Pads | |
530 mm | |
Concrete, Granite, Industrial Floors, Linoleum, Marble, Stoneware, Terrazzo, VCT, and Vinyl Flooring |
Brown | |
Cleaning and Polishing Floors | |
Round |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Scotch-Brite™ Clean & Shine Floor Pad > 530 mm
Spot an opportunity for improvement?Share a Content Correction