missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ ClearKlens Plus Detergent
Cleans in general purpose applications within various industries, including pharmaceutical and cosmetic. Diversey ClearKlens Plus Detergent is a concentrated, neutral, high-foaming liquid detergent.
Brand: Diversey™ 7513374
This item is not returnable.
View return policy
Description
- Concentrated, neutral, high-foaming liquid detergent
- Designed for general purpose applications
- Rinse after use to remove solutions from product contact surfaces
Specifications
Liquid | |
Non-sterile | |
Canister | |
2 Pk. |
∼6 | |
0.1 to 1% | |
Unfiltered | |
5 L |