missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CLED Agar

Product Code. 13245349
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
CLED Agar
Manufacturing Origin:
Germany
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 13245349

Brand: Thermo Scientific™ PO5009A

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Isolate, enumerate and presumptively identify all pathogens occurring in urinary tract infections with this traditional media which provides good colonial differentiation.

Detect common urinary tract pathogens using Thermo Scientific™ CLED Agar, a traditional media which provides good colonial differentiation, clear diagnostic characteristics and prevents swarming from Proteus spp. due to the electrolyte deficiency in the formulation.

Isolate, enumerate and presumptively identify all UTI related microorganisms with this traditional urinary bacteriology medium which provides a valuable non-inhibitory diagnostic for plate culture.

This product may not be available in all countries or territories. Please consult your local representative for availability.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale green, transparent
Content And Storage 2° to 12°C, in the dark
Format 90mm Monoplate
Manufacturing Origin Germany
Media Type CLED Agar
Product Type Agar
Fill Weight 17 g ± 5 %
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.