missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLIF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541601
This item is not returnable.
View return policy
Description
Peptide sequence: PTAMGSFSSH MTEFPRKRKG SDSDPSQSGI MTEKVVEKLS QNPLTYLLST Sequence homology: Dog: 86%; Human: 100%; Mouse: 77%.
This gene encodes a basic helix-loop-helix transcription factor belonging to the PAS (PER, ARNT, SIM) superfamily. The PAS proteins play important roles in adaptation to low atmospheric and cellular oxygen levels, exposure to certain environmental pollutants, and diurnal oscillations in light and temperature. This protein forms a transcriptionally active heterodimer with the circadian CLOCK protein, the structurally related MOP4, and hypoxia-inducible factors, such as HIF1alpha. Consistent with its role as a biologically relevant partner of circadian and hypoxia factors, this protein is coexpressed in regions of the brain such as the thalamus, hypothalamus, and amygdala. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Oct 2011].
Specifications
| CLIF | |
| Polyclonal | |
| Unconjugated | |
| ARNTL2 | |
| 4632430A05Rik; Arnt4; Arntl2; aryl hydrocarbon receptor nuclear translocator like 2; aryl hydrocarbon receptor nuclear translocator-like 2; aryl hydrocarbon receptor nuclear translocator-like protein 2; Basic-helix-loop-helix-PAS protein MOP9; BHLHE6; BMAL2; brain and muscle ARNT-like 2; brain and muscle Arnt-like protein 2 variant d; brain-muscle-ARNT-like protein 2; class E basic helix-loop-helix protein 6; CLIF; CYCLE-like factor; member of PAS protein 9; MGC149671; MGC149672; MOP9; PAS domain-containing protein 9; PASD9; transcription factor BMAL2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56938 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WYA1 | |
| ARNTL2 | |
| Synthetic peptide directed towards the N-terminal of human ARNTL2 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido