Learn More
Clinofibrate, MedChemExpress
Product Code. 16421897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16421897
Brand: Medchem Express HY13528
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.
Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.
Specifications
Chemical Name or Material Clinofibrate
CAS 30299-08-2
Color White
Purity Grade Notes Research
Molecular Formula C28H36O6
Quantity 50 mg
Synonym S-8527
Solubility Information DMSO : ≥ 30 mg/mL (64.02 mM)
SMILES CCC(OC1=CC=C(C2(C3=CC=C(OC(CC)(C)C(O)=O)C=C3)CCCCC2)C=C1)(C)C(O)=O
Molecular Weight (g/mol) 468.58
Formula Weight 468.58
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism