missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Coagulation Factor III/Tissue Factor Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
437.00 € - 1080.00 €
Specifications
Assay Range | 23.45 pg/mL |
---|---|
Assay Sensitivity | 23.45-1500 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18232226
|
Novus Biologicals™
NBP2-79650-15PLATES |
15 Plates |
1080.00 €
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18645010
|
Novus Biologicals™
NBP2-79650-5Plates |
5 Plates |
437.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Rat Coagulation Factor III/Tissue Factor.Specifications
23.45 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes an antibody specific for Rat Coagulation Factor III/Tissue Factor. | |
Sandwich ELISA | |
2152 | |
Rabbit/Mouse | |
Cancer | |
Coagulation Factor III/Tissue Factor |
23.45-1500 pg/mL | |
HRP | |
Antibody Pairs | |
Rat | |
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
F3 | |
RUO | |
Storage is content dependent. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Coagulation Factor III/Tissue Factor Antibody Pair [HRP]