Learn More
Hach Lange™ COD Cuvette Test
Click to view available options
Range:
150 to 1000 mg/L O2
Unit Size:
Each
Especificaciones
Especificaciones
| Certifications/Compliance | ISO 6060-1989, DIN 38409-H41-H44 |
| For Use With (Application) | For Automatic Recognition in the Photometer |
| No. of Tests | 25 |
| Range | 150 to 1000 mg/L O2 |
| Test Method | Dichromate |
| Type | COD Cuvette Test |
Seguridad y manipulación
Nombre del producto
- CSB/COD/DCO
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Respiratory sensitiser Category 1A
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- H290-May be corrosive to metals.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Sulphuric acid, Mercury(II) sulfate, Silver sulfate, potassium dichromate
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido