Learn More
KGW Isotherm™ Cold Trap
Click to view available options
Model:
KF 29-GL
KF 29-GL-A
KFL 29-GL
KFL 29-GL-A
Includes:
12C Dewar
12C Dewar, Spout
18C Dewar
18C Dewar, Spout
missing translation for 'unitSize'
Each
Specifications
Specifications
| Capacity (Metric) | 150 mL (Condensate), 1 L (Coolant) |
| Includes | 12C Dewar |
| Quantity | 1 Pack |
| Type | Cold Trap (Short) |
| Model | KF 29-GL |
| Connection Type | Glass Thread GL18 with Olive |