Learn More
VACUUBRAND™ Cold Trap
Click to view available options
Type:
GKF 1000i Cold Trap
SKF H 25 Cold Trap
SKF H 40 Cold Trap
Unit Size:
Each
Description
Cold trap SKF
Cold trap GKF
Inline oil filter HF
- Sturdy, easy to clean
- Easy to disassemble
- Two-wall design with good conductance
- Long operating time per coolant filling
- Easy condensate drainage without disassembling
Cold trap GKF
- Highly reflective insulation jacket for extended coolant life
- Vertical window: Direct observation of condensate and coolant levels
- PTFE stopcock: Drain condensate without disassembly
- Sheet metal shield for protection against external damage and implosion
Inline oil filter HF
- Extended oil lifetime
- Reduced service demands
- Easy space-saving mounting
- Simple filter change, service indicator
Specifica
Specifica
| Case Material | Borosilicate Glass, PTFE, FKM |
| For Use With (Equipment) | Vacuubrand pumps |
| Type | GKF 1000i Cold Trap |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto