missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Autoclavable Sanitary SIP Connectors

Product Code. 10360825
Click to view available options
Hose Barb:
1/2 Hose Barb
3/8 Hose Barb
Tubing Size:
3/4 x 1-1/2 in.
3/4 x 3/4 in.
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 10360825

Brand: CPC™ STC1700600

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance USP Class VI
For Use With (Application) SIP Processing
Tubing Size 3/4 x 3/4 in.
Hose Barb 3/8 Hose Barb
Material (Seal) Platinum-cured Silicone
Format Free-floating
Product Type ADCF SIP Connector
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.