Learn More
OHAUS™ Column Clamp
OHAUS™ Column Clamp is designed to securely grip and position a variety of laboratory glassware.
Brand: OHAUS™ 30392315
This item is not returnable.
View return policy
Description
- Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
- Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
- Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
Specifications
Column Clamp | |
Clamp, Specialty, Column, CLS-COLMNSM | |
12.41 in. | |
315.00 mm | |
Medium | |
Column |
Stainless Steel | |
11 mm | |
8.310 in. | |
211 mm | |
91 to 114 mm |